Order Entry
Netherlands
ContactUsLinkComponent
SBB Aqua 26 Plus, 26 litre unstirred boiling water bath. Temp range: 100C only. Mains supply: 230V AC 50/60Hz. Includes polycarbonate lid, base tray and UK cordset 1 * 1 items
Catalog # 462-0169
Supplier:  GRANT INSTRUMENTS
SBB Aqua 26 Plus, 26 litre unstirred boiling water bath. Temp range: 100C only. Mains supply: 230V AC 50/60Hz. Includes polycarbonate lid, base tray and UK cordset 1 * 1 items
Catalog # 462-0169
Supplier:  GRANT INSTRUMENTS
Frequently Bought Together