Order Entry
Netherlands
ContactUsLinkComponent
GLS Aqua 18 Plus, 18 litre Digital unstirred water Electronic PID controlled. Temp range: Ambient+5C to 99C. Temp stability at 37C: 0.1C. Shaking speed range: 40 to 400 strokes/min. Mains supply: 230V AC 50/60Hz. Includes ploycarbonate lid, bas 1 * 1 items
Catalog # 462-0157
Supplier:  GRANT INSTRUMENTS
GLS Aqua 18 Plus, 18 litre Digital unstirred water Electronic PID controlled. Temp range: Ambient+5C to 99C. Temp stability at 37C: 0.1C. Shaking speed range: 40 to 400 strokes/min. Mains supply: 230V AC 50/60Hz. Includes ploycarbonate lid, bas 1 * 1 items
Catalog # 462-0157
Supplier:  GRANT INSTRUMENTS
Frequently Bought Together