Order Entry
Netherlands
ContactUsLinkComponent
JB Aqua Dual Plus, 5 & 12 litre Dual Analogue unstirred water bath thermostatically controlled. Temp range: Ambient+5C to 98C. Temp stability at 37C: 1.0C. Mains supply: 230V AC 50/60Hz. Includes polycarbonate lid, base tray and UK cords 1 * 1 items
  462-0185
JB Aqua Dual Plus, 5 & 12 litre Dual Analogue unstirred water bath thermostatically controlled. Temp range: Ambient+5C to 98C. Temp stability at 37C: 1.0C. Mains supply: 230V AC 50/60Hz. Includes polycarbonate lid, base tray and UK cords 1 * 1 items
  462-0185
Frequently Bought Together