Tubing, Dual Layer PEEK, 1/16'' x 0.18 mm ID, solid yellow outside, natural inside, 10 m/pkg 1 * 10 m
Catalog # JOURJR-TD-6008-M10
Supplier: VICI JOUR RESEARCH
Tubing, Dual Layer PEEK, 1/16'' x 0.18 mm ID, solid yellow outside, natural inside, 10 m/pkg 1 * 10 m
Catalog # JOURJR-TD-6008-M10
Supplier: VICI JOUR RESEARCH
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection