ESHMUNO Q This product cannot be returned after delivery 1 * 10 mL
Catalog # 1.20079.0010
Supplier: MERCK PRODUCTION CHEMICALS
ESHMUNO Q This product cannot be returned after delivery 1 * 10 mL
Catalog # 1.20079.0010
Supplier: MERCK PRODUCTION CHEMICALS
Frequently Bought Together
o-Xylene anhydrous, 97 % 1 * 1 L
Supplier: Merck
n-Hexane ≥95%, TECHNICAL
Supplier: VWR Chemicals
Dimethyl sulphoxide, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Acetonitrile, ChromAR® for HPLC, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
n-Hexane, EMPLURA®, Supelco®
Supplier: Merck
Diethylene glycol monobutyl ether ≥99%
Supplier: Thermo Fisher Scientific
Diisopropyl ether 98+% stabilised, pure
Supplier: Thermo Fisher Scientific
Water ACS, Deionized
Supplier: Thermo Fisher Scientific
1,2-Dimethoxyethane ≥99% inhibitor-free, Reagent Grade
Supplier: Honeywell Chemicals
Glycerine, anhydrous ≥99.5% (by GC), Puriss. p.a. ACS, distilled, Riedel-de Haën™
Supplier: Honeywell Chemicals
Butyl acetate 99+%, extra pure
Supplier: Thermo Fisher Scientific
n-Pentane, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific