Order Entry
Netherlands
ContactUsLinkComponent
Hydrometer, ISO 650, series L50, range 0.850-0.900, length 335mm, accuracy 0.0005, div 0,0005g/ml, calibration temperature 60/60 F. 1 * 1 items
Catalog # ALLA3502LC08520BQP
Supplier:  Alla France
Hydrometer, ISO 650, series L50, range 0.850-0.900, length 335mm, accuracy 0.0005, div 0,0005g/ml, calibration temperature 60/60 F. 1 * 1 items
Catalog # ALLA3502LC08520BQP
Supplier:  Alla France
Frequently Bought Together