SWAB WITH 10ML BUFFERED PEPTONIC WATER 1 * 40 Pce
Catalog # TRITB601.23.0010
Supplier: TRITIUM
Frequently Bought Together
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Test strips, peracetic acid, Dosatest
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Aluminium foils rolls and dispenser
Supplier: Korff
VWR®, Microscope Slides
Supplier: VWR Collection
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
BD Plastipak™ Syringes
Supplier: BD Medical
Sulphuric acid 25%, GPR RECTAPUR®
Supplier: VWR Chemicals
Dip slides for surfaces and in liquids, Envirocheck®
Supplier: Merck
Hydrochloric acid 0.1 mol/l (0.1 N), AVS TITRINORM Reag. Ph. Eur. volumetric solution
Supplier: VWR Chemicals
VWR®, Zip-Seal Bags
Supplier: VWR Collection
Accessories
Supplier: Merck
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection