Spare PTFE Keys for Stopcocks, PYREX®
Spare PTFE keys for stopcocks, PYREX®, straight bore, 2,5 mm
Catalog # BIBBSST103K
Supplier: SciLabware
Jump to:
Product Details & Documents
Fully interchangeable PTFE keys for Pyrex® stopcocks.
- Supplied complete with plastic screw thread retaining device
- Self lubricating, avoiding the need for stopcock grease
- Made to a taper of 1:10 giving a combination of performance and ease of operation
Leveringsinformatie: Key is supplied complete with plastic screw thread retaining device.
Specifications
- Hole Ø:2,5 mm
- Description:Spare PTFE keys for stopcocks, PYREX®, straight bore
- Seq:02