Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Dexamethasone ≥98%

Dexamethasone ≥98%

Glass bottle for solids, 1 g
Catalog # A17590.03
Health
Dexamethasone ≥98%
Catalog # A17590.03
CAS Number:  50-02-2
Dexamethasone ≥98%
Dexamethasone ≥98%

Glass bottle for solids, 1 g

CAS Number:  50-02-2
Price
167,00 €
Log in for availability
1 * 1 g
Product Details & Documents

Purity: 98% CAS No.: 50-02-2 Molecular Formula: C22H29FO5 Formula Weight: 392.47 Melting Point: ca 260° dec. Beilstein Reference: 2066651 Merck Reference: 14,2943 Optical Rotation: +75° (c=1 in doxane) MDL No.: MFCD00064136

Anti-inflammatory glucocorticoid

Synonyms:  Dexamethasonum, 9-Fluor-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadien-3,20-dion
  • CAS Number:50-02-2
  • Formula:C₂₂H₂₉FO₅
  • MDL Number:MFCD00064136
  • Molecular Weight:392.47 g/mol
  • Merck Index:14,2943
  • Melting Pt:∼ 260 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:200-003-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.