Order Entry
Netherlands
ContactUsLinkComponent
Insert, volume 5mm, colour clear, material glass, usable volume 210µl, residual volume <3µl, conical base, used inside 8x40mm flat b ottom shell vial, polySpring provides a cushion against needle contact
Catalog # CRMAC4015-96A
Supplier:  CHROMACOL
Insert, volume 5mm, colour clear, material glass, usable volume 210µl, residual volume <3µl, conical base, used inside 8x40mm flat b ottom shell vial, polySpring provides a cushion against needle contact
Catalog # CRMAC4015-96A
Supplier:  CHROMACOL