Order Entry
Netherlands
ContactUsLinkComponent
Insert, volume 6mm, flanged lip, material glass, colour clear, mandrel tip, usable volume 500µl, residual volume <6µl, self aligning and provide a cushion against needle contact, flange improves centering of lip
Catalog # CRMAC4015-641
Supplier:  CHROMACOL
Insert, volume 6mm, flanged lip, material glass, colour clear, mandrel tip, usable volume 500µl, residual volume <6µl, self aligning and provide a cushion against needle contact, flange improves centering of lip
Catalog # CRMAC4015-641
Supplier:  CHROMACOL