Order Entry
Netherlands
ContactUsLinkComponent
Closure, cap size 11mm, cap colour blue, target snap it, septum clear PTFE/red rubber, natural rubber septum is moderately resealabl e, compatible with many GC applications
Catalog # CRMAC4011-51B
Supplier:  CHROMACOL
Closure, cap size 11mm, cap colour blue, target snap it, septum clear PTFE/red rubber, natural rubber septum is moderately resealabl e, compatible with many GC applications
Catalog # CRMAC4011-51B
Supplier:  CHROMACOL