Order Entry
Netherlands
ContactUsLinkComponent
Closure, cap size 11mm, cap colour blue, target snap it, septum integral molded in polyethylene, single use economy cap, not reseala ble, do not use with concentrated aromatic hydrocarbons
Catalog # CRMAC4011-50B
Supplier:  CHROMACOL
Closure, cap size 11mm, cap colour blue, target snap it, septum integral molded in polyethylene, single use economy cap, not reseala ble, do not use with concentrated aromatic hydrocarbons
Catalog # CRMAC4011-50B
Supplier:  CHROMACOL