Order Entry
Netherlands
ContactUsLinkComponent
Vial, capacity 2ml, target snap-it, 11mm crimp/snap, clear, glass I-D™, flat bottom, diameter 12mm, height 32mm, usable volume 1.5ml , residual volume <170µl, patched
Catalog # CRMAC4011-5W
Supplier:  CHROMACOL
Vial, capacity 2ml, target snap-it, 11mm crimp/snap, clear, glass I-D™, flat bottom, diameter 12mm, height 32mm, usable volume 1.5ml , residual volume <170µl, patched
Catalog # CRMAC4011-5W
Supplier:  CHROMACOL