Order Entry
Netherlands
ContactUsLinkComponent
Insert, volume 6mm, colour clear, material glass, precision mandrel tip, allows for the greatest sample withdrawal
Catalog # CRMAC4010-629L
Supplier:  CHROMACOL
Insert, volume 6mm, colour clear, material glass, precision mandrel tip, allows for the greatest sample withdrawal
Catalog # CRMAC4010-629L
Supplier:  CHROMACOL