Order Entry
Netherlands
ContactUsLinkComponent
qScript One-Step RT and 4x concentrated master mix optimized for MGB probes contains all components except gene-specific primers and template for fast one-step real-time qRT-PCR on Applied Biosystems 7000, 7300, 7700, or 7900 and StepOne# instrument 1 * 100 Assays
Catalog # 733-1908
Supplier:  Quantabio
qScript One-Step RT and 4x concentrated master mix optimized for MGB probes contains all components except gene-specific primers and template for fast one-step real-time qRT-PCR on Applied Biosystems 7000, 7300, 7700, or 7900 and StepOne# instrument 1 * 100 Assays
Catalog # 733-1908
Supplier:  Quantabio
Frequently Bought Together