Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Accessories for connection system GLS 80
Accessories for connection system GLS 80
O-ring gasket seal from EPDM for GLS 80 bottles, I.Ø. 63,5 mm, O.Ø. 78 mm
Catalogus # SCOT1152913
Leverancier:   DWK Life Sciences
Accessories for connection system GLS 80
Catalogus # SCOT1152913
Leverancier:  DWK Life Sciences
Accessories for connection system GLS 80
Accessories for connection system GLS 80
O-ring gasket seal from EPDM for GLS 80 bottles, I.Ø. 63,5 mm, O.Ø. 78 mm
Price
2,40 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Specificaties
  • Description:
    O-ring gasket seal from EPDM for GLS 80 bottles, I.Ø. 63,5 mm, O.Ø. 78 mm
  • Seq:
    09