Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Sample Containers

Sample Containers

250 ml, 82×108×49 mm
Catalog # 216-4737
Supplier:   BRABO VERPAKKINGEN
Sample Containers
Catalog # 216-4737
Supplier:  BRABO VERPAKKINGEN
Sample Containers
Sample Containers

250 ml, 82×108×49 mm

Price
103,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

PS, clear.

  • Inexpensive, single-use
  • Optimum content visibility
  • Optional matching lids available

Universal container for a wide variety of uses. Ideal for collecting, storing and transporting samples.

Specifications
  • Capacity:
    250 ml
  • W×D×H:
    82×108×49 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.