Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Vials with insert, WHEATON®

Vials with insert, WHEATON®

Vials with insert, 0,50 ml, 12×32 mm, No, Snap/crimp
Catalog # WHEAW225187
Supplier:   DWK Life Sciences
Vials with insert, WHEATON®
Catalog # WHEAW225187
Supplier:  DWK Life Sciences
Vials with insert, WHEATON®
Vials with insert, WHEATON®

Vials with insert, 0,50 ml, 12×32 mm, No, Snap/crimp

Price
353,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

These vials made of PP uses either aluminum crimp seals or plastic snap cap closures, which eliminate the need for crimping and decapping tools, and provide a larger target area.

  • Available with snap/crimp or screw top finish
  • Unique cone-shaped bottom ensures maximum sample retrieval

Packaging: Packaged as 100 vials per shelf pack or 1000 vials per case.

Specifications
  • Cap included:
    No
  • Ø×H:
    12×32 mm
  • Capacity:
    0,50 ml
  • Description:
    Vials with insert
  • Cap type:
    Snap/crimp