Fittings
Gripper nut, PEEK, M6, for 1/16" tubing
Catalogus # OMNI002135
Leverancier: OMNIFIT
Ga naar:
Product Details & Documents
Documents
Certificates
Zoeken op lotnummerSpecificaties
- Description:Gripper nut, PEEK, M6, for 1/16" tubing
- Seq:40