Order Entry
Netherlands
ContactUsLinkComponent
VYCOR standard wall tubing has an O.D. of 13mm and is 914mm (36 ) long, approximately 6mm ( ) with cut ends. Material: 96% Silica Glass Code 7913 Wall Thickness: 1.2 mm (approx) Wall Limits: .041-.053 inches O.D. Limits: .496-.528 mm Nominal Size 1 * 10 items
Catalog # BIBBS/8743173
Supplier:  SciLabware
VYCOR standard wall tubing has an O.D. of 13mm and is 914mm (36 ) long, approximately 6mm ( ) with cut ends. Material: 96% Silica Glass Code 7913 Wall Thickness: 1.2 mm (approx) Wall Limits: .041-.053 inches O.D. Limits: .496-.528 mm Nominal Size 1 * 10 items
Catalog # BIBBS/8743173
Supplier:  SciLabware
Frequently Bought Together