Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

n-Dodecane, Supelco®

Glass bottle, 5 mL
Catalog # 44010-5ML
Health
Supplier:   Merck
n-Dodecane, Supelco®
Catalog # 44010-5ML
Supplier:  Merck
CAS Number:  112-40-3
n-Dodecane, Supelco®
n-Dodecane, Supelco®

Glass bottle, 5 mL

CAS Number:  112-40-3
Price
70,60 €
Log in for availability
1 * 5 mL
Product Details & Documents
Synonyms:  Dodecane
  • CAS Number:112-40-3
  • Formula:H₃C(CH₂)₁₀CH₃
  • MDL Number:MFCD00008969
  • Molecular Weight:170.34 g/mol
  • Boiling Pt:216 °C (1013 hPa)
  • Melting Pt:–9.6 °C
  • Density:0.7487 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ100
  • Flash Pt:74 °C
  • Storage Temperature:Ambient
  • EINECS:203-967-9
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL

Specifications