16 EPA PRIORITY PAH POLLUTANTS MIXTU 1 * 4,5 mL
Catalog # PROODE-PROM16
Supplier: LGC Standards PROMOCHEM
16 EPA PRIORITY PAH POLLUTANTS MIXTU 1 * 4,5 mL
Catalog # PROODE-PROM16
Supplier: LGC Standards PROMOCHEM
Frequently Bought Together
Glycerine (vegetable based), anhydrous, EMPROVE® EXPERT ACS, Ph. Eur., BP, USP, JP for biopharmaceutical production, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Glycerine ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Dichloromethane, dehydrated (max. 0.01% H₂O) ≥99% stabilised, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
2-Propanol, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
ACETONITRILE EMPLURA® 1 * 4 L
Supplier: Merck
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
1-Propanol, anhydrous ≥99.9%
Supplier: Thermo Fisher Scientific
Tetrahydrofuran, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Cyclohexane 99+% ACS, meets analytical specifications of Reagent Ph. Eur. for spectroscopy
Supplier: Thermo Fisher Scientific
Carbon disulphide 99.9% for HPLC
Supplier: Thermo Fisher Scientific