BISPHENOL A (UNLABELLED) 100 UG/ML IN AC 1 * 1,2 mL
PROOCILULM71061.2
Frequently Bought Together
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
Ethylenediamine 99%
: Thermo Fisher Scientific
n-Pentane 98%, Reagent Grade
: Honeywell Chemicals
Bis(2-methoxyethyl)ether, extra dry over Molecular Sieve 99+%, AcroSeal®
: Thermo Fisher Scientific
Acetone ≥99.5% (by GC), liquid USP, BP, Ph. Eur., pure
: PanReac AppliChem
Dimethyl sulphoxide, extra dry over Molecular Sieve 99.7+%, AcroSeal®
: Thermo Fisher Scientific
Acetonitrile 99.9%, ECD tested for residue analysis
: Thermo Fisher Scientific
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
: PanReac AppliChem
Dimethyl sulphoxide, anhydrous ≥99.8%
: Thermo Fisher Scientific