LYSING MATRIX B 50x2ml tubes 1 * 50 items
Catalog # ICNA6911050
Supplier: MP Biomedicals
Frequently Bought Together
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Pipette tips, Standard
Supplier: Brand
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98%, TECHNICAL
Supplier: VWR Chemicals
Protein isolation and purification, FastPROTEIN™ blue matrix kit, MP Biomedicals
Supplier: MP Biomedicals
Conical separating funnels
Supplier: Lenz Laborglas GmbH & CO.KG
DETA NONOate (NOC-18) 95%
Supplier: Thermo Fisher Scientific
10% SDS (Sodium Dodecyl Sulfate), Corning®
Supplier: Corning