Boranepyridine complex 95%
Glass bottle, 100 mL
Supplier: Thermo Fisher Scientific
Boranepyridine complex 95%
Catalog # ACRO177011000
Supplier: Thermo Fisher Scientific
CAS Number: 110-51-0
Jump to:
Product Details & Documents
Synonyms: Pyridine Borane, Borane, pyridine complex
- CAS Number:110-51-0
- Formula:C₅H₈BN
- MDL Number:MFCD00012435
- Molecular Weight:92.94 g/mol
- Melting Pt:10…11 °C
- Density:0.92 g/cm³
- Flash Pt:73 °C
- Storage Temperature:Refrigerator
- UN:2810
- ADR:6.1,II
Documents
ACRO17701
SDS
Specifications
Specification Test Results
- Appearance:Clear to slightly hazy colorless to amber Liquid (May darken on storage)
- Infrared spectrum:Conforms
- Titration:≥94 % (Active hydrogen)
- HPLC:≥95.0 %
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Cat. No.:ACRO177011000



