Diethyl ether, extra dry 99.5% stabilised, AcroSeal®
Glass bottle, 1 L
Supplier: Thermo Fisher Scientific
Diethyl ether, extra dry 99.5% stabilised, AcroSeal®
Catalog # 326860010.
Supplier: Thermo Fisher Scientific
CAS Number: 60-29-7
Jump to:
Product Details & Documents
Synonyms: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
- CAS Number:60-29-7
- Formula:(CH₃CH₂)₂O
- MDL Number:MFCD00011646
- Molecular Weight:74.12 g/mol
- Merck Index:12,03852
- Boiling Pt:34.6 °C (760 mmHg)
- Melting Pt:–116 °C
- Density:0.714 g/cm³
- Flash Pt:–45 °C
- Storage Temperature:Ambient
- EINECS:200-467-2
- UN:1155
- ADR:3,I
- REACH:01-2119535785-29
Documents
Specifications
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.005 % (Coulometric)
- GC:≥99.4 %
- Stabilizer:1.5 to 10 ppm BHT
- Residue after evaporation:≤0.001 %
- Acidity:≤0.0002 meq/g
- Filt.over 0.2 micron PFTE:Filtered
- Peroxides:≤0.002 % (as H2O2)
Specifications
- Pack type:Glass bottle
- Pk:1 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



