Nipecotamide 95%
Glass bottle, 50 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Piperidine-3-carboxamide
- CAS Number:4138-26-5
- Formula:C₆H₁₂N₂O
- MDL Number:MFCD00005993
- Molecular Weight:128.1700 g/mol
- Melting Pt:102…108 °C
- Storage Temperature:Ambient
- EINECS:223-962-5
Documents
ACRO12830
SDS
Specifications
Specification Test Results
- Appearance:Yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:102 to 108 °C
- Titration with HClO4:≥94 %
Specifications
- Pack type:Glass bottle
- Pk:50 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



