[DE]MO A ENTEROVIRUS CLONE 5-D81 (1 ML) 1 * 1 ST
Catalog # OXOIM706401-2
Supplier: THERMO FISHER DIAGNOSTICS
[DE]MO A ENTEROVIRUS CLONE 5-D81 (1 ML) 1 * 1 ST
Catalog # OXOIM706401-2
Supplier: THERMO FISHER DIAGNOSTICS
Frequently Bought Together
Aceton ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Nitril handschoenen, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR®, Serologische pipetten, standaardreeks
Supplier: VWR Collection
VWR®, Transferpipetten, Hoge Prestaties
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrilhandschoenen
Supplier: VWR Collection
VWR®, Nitrilhandschoenen
Supplier: VWR Collection
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
pH-teststrips, MQuant®, Supelco®
Supplier: Merck
VWR®, gezichtstissues
Supplier: VWR Collection