Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Biotin conjugation estimation, HOOK™ BiotinQuant
Biotin conjugation estimation, HOOK™ BiotinQuant
HOOK™ BiotinQuant Kit, 1 KIT
Catalog # BKC-01
Supplier:   G-Biosciences
Biotin conjugation estimation, HOOK™ BiotinQuant
Catalog # BKC-01
Supplier:  G-Biosciences
CAS Number:  
Biotin conjugation estimation, HOOK™ BiotinQuant
Biotin conjugation estimation, HOOK™ BiotinQuant
HOOK™ BiotinQuant Kit, 1 KIT
CAS Number:  
Price
273,00 €
Log in for availability
1 * 1 Kit
Product Details & Documents
HOOK™ BiotinQuant is designed for the estimation of biotin conjugation. It measures biotin using HABA [4’-hydroxyazobenzene-2-carboxylic acid] dye. HABA binds with avidin at the biotin-binding site.
Documents
Specifications
Specifications
  • Pk:
    1 KIT
  • Description:
    HOOK™ BiotinQuant Kit