Order Entry
Netherlands
ContactUsLinkComponent
A high salt RAB (Reassembly) buffer for the lysis of mammalian cells. Designed for the extraction of soluble proteins, does not extract detergent extractable insoluble proteins. 1 * 250 mL
Catalog # GENO786-91
Supplier:  G-Biosciences
A high salt RAB (Reassembly) buffer for the lysis of mammalian cells. Designed for the extraction of soluble proteins, does not extract detergent extractable insoluble proteins. 1 * 250 mL
Catalog # GENO786-91
Supplier:  G-Biosciences
Frequently Bought Together