Order Entry
Netherlands
ContactUsLinkComponent
Fully automatic, microprocessor-controlled vertical laboratory autoclave with Cooling Sytem and CH-plug. Chamberdimension (xL) 500 x 750 mm Volume: 160 liters 1 * 1 items
Catalog # 481-2155
Supplier:  TUTTNAUER
Fully automatic, microprocessor-controlled vertical laboratory autoclave with Cooling Sytem and CH-plug. Chamberdimension (xL) 500 x 750 mm Volume: 160 liters 1 * 1 items
Catalog # 481-2155
Supplier:  TUTTNAUER
Frequently Bought Together