Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
REFRACTIVE INDEX LIQUID SERIES A REFRACTIVE INDEX @ 25°C: 1. 55600 1 * 1 ST
Catalog # CARG1809X
Supplier:
CARGILLE
Expand image
REFRACTIVE INDEX LIQUID SERIES A REFRACTIVE INDEX @ 25°C: 1. 55600 1 * 1 ST
Catalog # CARG1809X
Supplier:
CARGILLE
Frequently Bought Together
Test kits, digital titration method
Supplier: Hach
SAMPLE CHAMBER CLOSURE 1 * 12 items
Supplier: Epredia
Colorimetric Rapid Tests, Aluminium, VISOCOLOR®
Supplier: MACHEREY-NAGEL
Potassium hydroxide 45% in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier: Avantor