Order Entry
Netherlands
ContactUsLinkComponent
Silent aircompresor for autoclaves with rapid cooling. Capacity 30 l/min, for models 2540-, 3150 ELC and ELVC 1 * 1 items
Catalog # 481-2086
Supplier:  TUTTNAUER
Silent aircompresor for autoclaves with rapid cooling. Capacity 30 l/min, for models 2540-, 3150 ELC and ELVC 1 * 1 items
Catalog # 481-2086
Supplier:  TUTTNAUER
Frequently Bought Together