Order Entry
Netherlands
ContactUsLinkComponent
Fully automatic, microprocessor-controlled benchtop laboratory autoclave with CH-plugChamberdimension (xL) 310 x 500 mm Volume: 40 liters 1 * 1 items
Catalog # 481-2020
Supplier:  TUTTNAUER
Fully automatic, microprocessor-controlled benchtop laboratory autoclave with CH-plugChamberdimension (xL) 310 x 500 mm Volume: 40 liters 1 * 1 items
Catalog # 481-2020
Supplier:  TUTTNAUER
Frequently Bought Together