Clamps with M10 thread, flexible
Clamp, round jaws, 230 g, 12 - 25 mm
Catalogus # USBE2686
Leverancier: USBECK Laborgeräte
Ga naar:
Product Details & Documents
This flexible clamp is ideal for use with retort stand bases and lab jacks.
- With M10 thread
- Three-pronged or with round jaws
Specificaties
- Spanning width:12 - 25 mm
- Description:Clamp, round jaws, 230 g
Aanbevelingen
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



