Pipette, 50:0.2ml, graduated, class AS, DIN 12697 ISO product with single certificate 1 * 1 items
Catalog # HIRS110017508
Supplier: HIRSCHMANN
Pipette, 50:0.2ml, graduated, class AS, DIN 12697 ISO product with single certificate 1 * 1 items
Catalog # HIRS110017508
Supplier: HIRSCHMANN
Frequently Bought Together
Volumetric flasks, USP, class A, blue markings, BLAUBRAND®
Supplier: Brand
VWR® Measuring Cylinders, Low Form, Class B, Borosilicate Glass
Supplier: VWR Collection
Measuring cylinders, Tall form, class A, borosilicate glass, Schellbach Stripe
Supplier: HIRSCHMANN
Bulb pipettes, BLAUBRAND®, two marks, class AS
Supplier: Brand
VWR® Volumetric Flasks, Class A, Glass, Clear, NS Plastic Stopper
Supplier: VWR Collection
Volumetric flasks, class A, blue markings, BLAUBRAND®
Supplier: Brand
VWR® Burettes, with PTFE Key Stopcock
Supplier: VWR Collection
VWR® Measuring Cylinders, Tall Form, Class A, Borosilicate Glass
Supplier: VWR Collection
Volumetric flasks, class A, blue markings, BLAUBRAND®
Supplier: Brand
VWR®, Hydrometers, M50
Supplier: VWR Collection
Volumetric flasks, class A, white markings
Supplier: witeg Labortechnik
Commercial grade hydrometers
Supplier: Alla France
Mixing measuring cylinders, tall form, class A, borosilicate glass
Supplier: HIRSCHMANN