Order Entry
Netherlands
Orders LinkContactUsLinkComponent
n-Dodecane ≥99%

n-Dodecane ≥99%

Glass bottle, 100 mL
Catalog # A14834.AE
Health
n-Dodecane ≥99%
Catalog # A14834.AE
CAS Number:  112-40-3
n-Dodecane ≥99%
n-Dodecane ≥99%

Glass bottle, 100 mL

CAS Number:  112-40-3
Price
39,30 €
Log in for availability
1 * 100 mL
Product Details & Documents

Purity: 99+% CAS No.: 112-40-3 Molecular Formula: CH3(CH2)10CH3 Formula Weight: 170.34 Flash Point: 71°(159°F) Physical Form: Liquid Density: 0.749 Refractive Index: 1.4220 Odour: Petrol-like Melting Point: -10° Boiling Point: 215-217° Beilstein Reference: 1697175 MDL No.: MFCD00008969

Synonyms:  Dodecane
  • CAS Number:112-40-3
  • Formula:H₃C(CH₂)₁₀CH₃
  • MDL Number:MFCD00008969
  • Molecular Weight:170.34 g/mol
  • Boiling Pt:215…217 °C
  • Melting Pt:–10 °C
  • Density:0.749
  • Flash Pt:71 °C (159 °F)
  • Storage Temperature:Ambient
  • EINECS:203-967-9
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.