n-Dodecane ≥99%
Glass bottle, 2.500 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99+% CAS No.: 112-40-3 Molecular Formula: CH3(CH2)10CH3 Formula Weight: 170.34 Flash Point: 71°(159°F) Physical Form: Liquid Density: 0.749 Refractive Index: 1.4220 Odour: Petrol-like Melting Point: -10° Boiling Point: 215-217° Beilstein Reference: 1697175 MDL No.: MFCD00008969
Synonyms: Dodecane
- CAS Number:112-40-3
- Formula:H₃C(CH₂)₁₀CH₃
- MDL Number:MFCD00008969
- Molecular Weight:170.34 g/mol
- Boiling Pt:215…217 °C
- Melting Pt:–10 °C
- Density:0.749
- Flash Pt:71 °C (159 °F)
- Storage Temperature:Ambient
- EINECS:203-967-9
Specifications
Specifications
- Pack type:Glass bottle
- Pk:2.500 mL



