D-(-)-Glutamic acid ≥99%
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
D-(-)-Glutamic acid ≥99%
Catalog # A14191.22
Supplier: Thermo Fisher Scientific
CAS Number: 6893-26-1
Jump to:
Product Details & Documents
Purity: 99+% CAS No.: 6893-26-1 Molecular Formula: C5H9NO4 Formula Weight: 147.13 Melting Point: 199° dec. Beilstein Reference: 1723800 Merck Reference: 14,4469 Optical Rotation: -30° (c=10 in 2N HCl) MDL No.: MFCD00063112
Synonyms: (R)-2-Aminopentanedioic acid, (R)-(-)-Glutamic acid, (R)-(-)-2-Aminopentanedioic acid, D(-)-2-Aminopentanedioic acid, D(-)-α-Glutamic acid, D(-)-1-Aminopropane-1,3-dicarboxylic acid, H-D-Glu-OH, (R)-(-)-α-Glutamic acid, (R)-(-)-1-Aminopropane-1,3-dicarboxylic acid
- CAS Number:6893-26-1
- Formula:C₅H₉NO₄
- MDL Number:MFCD00063112
- Molecular Weight:147.13 g/mol
- Merck Index:14,4469
- Boiling Pt:325 °C (1013 hPa)
- Melting Pt:199 °C (dec.)
- Density:1.533 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:230-000-8
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



