Order Entry
Netherlands
Orders LinkContactUsLinkComponent
D-(-)-Glutamic acid ≥99%

D-(-)-Glutamic acid ≥99%

Glass bottle for solids, 5 g
Catalogus # A14191.06
Leverancier:   Thermo Fisher Scientific
D-(-)-Glutamic acid ≥99%
Catalogus # A14191.06
CAS-nummer:  6893-26-1
D-(-)-Glutamic acid ≥99%
D-(-)-Glutamic acid ≥99%

Glass bottle for solids, 5 g

CAS-nummer:  6893-26-1
Price
48,80 €
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 99+% CAS No.: 6893-26-1 Molecular Formula: C5H9NO4 Formula Weight: 147.13 Melting Point: 199° dec. Beilstein Reference: 1723800 Merck Reference: 14,4469 Optical Rotation: -30° (c=10 in 2N HCl) MDL No.: MFCD00063112

Synonyms:  (R)-2-Aminopentanedioic acid, (R)-(-)-Glutamic acid, (R)-(-)-2-Aminopentanedioic acid, D(-)-2-Aminopentanedioic acid, D(-)-α-Glutamic acid, D(-)-1-Aminopropane-1,3-dicarboxylic acid, H-D-Glu-OH, (R)-(-)-α-Glutamic acid, (R)-(-)-1-Aminopropane-1,3-dicarboxylic acid
  • CAS Number:6893-26-1
  • Formula:C₅H₉NO₄
  • MDL Number:MFCD00063112
  • Molecular Weight:147.13 g/mol
  • Merck Index:14,4469
  • Boiling Pt:325 °C (1013 hPa)
  • Melting Pt:199 °C (dec.)
  • Density:1.533 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:230-000-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.