Order Entry
Netherlands
Orders LinkContactUsLinkComponent
2,4-Difluoroaniline ≥99%

2,4-Difluoroaniline ≥99%

Glass bottle for solids, 500 g
Catalog # A13673.36
Toxic
Irritant
2,4-Difluoroaniline ≥99%
Catalog # A13673.36
CAS Number:  367-25-9
2,4-Difluoroaniline ≥99%
2,4-Difluoroaniline ≥99%

Glass bottle for solids, 500 g

CAS Number:  367-25-9
Price
317,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 99% CAS No.: 367-25-9 Molecular Formula: C6H5F2N Formula Weight: 129.11 Flash Point: 62°(143°F) Refractive Index: 1.5070 Odour: Characteristic Melting Point: -8 to -7° Boiling Point: 168-170° Beilstein Reference: 2802556 Merck Reference: 14,3147 UN No.: 2941 MDL No.: MFCD00007648

  • CAS Number:367-25-9
  • Formula:C₆H₅F₂N
  • MDL Number:MFCD00007648
  • Molecular Weight:129.11 g/mol
  • Merck Index:14,3147
  • Boiling Pt:168…170 °C
  • Melting Pt:–8 °C…–7 °C
  • Melting Pt:–8 °C to –7 °C
  • Density:1.281
  • Flash Pt:62 °C (143 °F)
  • Storage Temperature:Ambient
  • EINECS:206-687-5
  • UN:2941
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications