Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

1,1,1-Tris(hydroxymethyl)ethane ≥97%

Glass bottle for solids, 2.500 g
Catalog # B24324.0E
1,1,1-Tris(hydroxymethyl)ethane ≥97%
Catalog # B24324.0E
CAS Number:  77-85-0
1,1,1-Tris(hydroxymethyl)ethane ≥97%
1,1,1-Tris(hydroxymethyl)ethane ≥97%

Glass bottle for solids, 2.500 g

CAS Number:  77-85-0
Price
127,00 €
Log in for availability
1 * 2.500 g
Product Details & Documents

Purity: 97% CAS No.: 77-85-0 Molecular Formula: C5H12O3 Formula Weight: 120.15 Flash Point: 160°(320°F) Melting Point: ca 190° Boiling Point: 137°/15mm Beilstein Reference: 1304452 MDL No.: MFCD00004687

Synonyms:  2-(Hydroxymethyl)-2-methyl-1,3-propanediol, Ethylidynetrimethanol
  • CAS Number:77-85-0
  • Formula:C₅H₁₂O₃
  • MDL Number:MFCD00004687
  • Molecular Weight:120.15 g/mol
  • Boiling Pt:137 °C (15 mmHg)
  • Melting Pt:∼ 190 °C
  • Flash Pt:160 °C (320 °F)
  • Storage Temperature:Ambient
  • EINECS:201-063-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2.500 g

Specifications