Order Entry
Netherlands
Orders LinkContactUsLinkComponent
D-(-)-Histidine ≥99%

D-(-)-Histidine ≥99%

Glass bottle for solids, 100 g
Catalog # B21027.22
D-(-)-Histidine ≥99%
Catalog # B21027.22
CAS Number:  351-50-8
D-(-)-Histidine ≥99%
D-(-)-Histidine ≥99%

Glass bottle for solids, 100 g

CAS Number:  351-50-8
Price
1 030,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 99% CAS No.: 351-50-8 Molecular Formula: C6H9N3O2 Formula Weight: 155.16 Melting Point: ca 285° dec. Beilstein Reference: 84089

Synonyms:  (R)-(-)-Histidine, D-3-Imidazol-4-ylalanine, (2R)-(-)-Histidine, H-D-His-OH
  • CAS Number:351-50-8
  • Formula:C₆H₉N₃O₂
  • MDL Number:MFCD00065963
  • Molecular Weight:155.16 g/mol
  • Merck Index:12,04758
  • Melting Pt:∼ 285 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:206-513-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications