Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

D-(-)-Histidine ≥99%

Glass bottle for solids, 25 g
Catalog # B21027.14
D-(-)-Histidine ≥99%
Catalog # B21027.14
CAS Number:  351-50-8
D-(-)-Histidine ≥99%
D-(-)-Histidine ≥99%

Glass bottle for solids, 25 g

CAS Number:  351-50-8
Price
281,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 99% CAS No.: 351-50-8 Molecular Formula: C6H9N3O2 Formula Weight: 155.16 Melting Point: ca 285° dec. Beilstein Reference: 84089

Synonyms:  (R)-(-)-Histidine, D-3-Imidazol-4-ylalanine, (2R)-(-)-Histidine, H-D-His-OH
  • CAS Number:351-50-8
  • Formula:C₆H₉N₃O₂
  • MDL Number:MFCD00065963
  • Molecular Weight:155.16 g/mol
  • Merck Index:12,04758
  • Melting Pt:∼ 285 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:206-513-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.