Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Propionic anhydride ≥97%

Propionic anhydride ≥97%

Glass bottle for solids, 500 g
Catalog # A12955.36
Corrosive
Propionic anhydride ≥97%
Catalog # A12955.36
CAS Number:  123-62-6
Propionic anhydride ≥97%
Propionic anhydride ≥97%

Glass bottle for solids, 500 g

CAS Number:  123-62-6
Price
39,50 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 97% CAS No.: 123-62-6 Molecular Formula: C6H10O3 Formula Weight: 130.14 Flash Point: 63°(145°F) Physical Form: Clear liquid Density: 1.012 Refractive Index: 1.4040 Sensitivity: Moisture Sensitive Appearance: Colourless Odour: Pungent Melting Point: -45° Boiling Point: 166-168° Beilstein Reference: 507066 Merck Reference: 14,7826 UN No.: 2496 MDL No.: MFCD00009303

  • CAS Number:123-62-6
  • Formula:C₆H₁₀O₃
  • MDL Number:MFCD00009303
  • Molecular Weight:130.14 g/mol
  • Merck Index:14,7826
  • Boiling Pt:166…168 °C
  • Melting Pt:–45 °C
  • Density:1.012
  • Flash Pt:63 °C (145 °F)
  • Storage Temperature:Ambient
  • EINECS:204-638-2
  • UN:2496
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.