Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

Propionic anhydride ≥97%

Glass bottle for solids, 250 g
Catalog # A12955.30
Corrosive
Propionic anhydride ≥97%
Catalog # A12955.30
CAS Number:  123-62-6
Propionic anhydride ≥97%
Propionic anhydride ≥97%

Glass bottle for solids, 250 g

CAS Number:  123-62-6
Price
27,90 €
Log in for availability
1 * 250 g
Product Details & Documents

Purity: 97% CAS No.: 123-62-6 Molecular Formula: C6H10O3 Formula Weight: 130.14 Flash Point: 63°(145°F) Physical Form: Clear liquid Density: 1.012 Refractive Index: 1.4040 Sensitivity: Moisture Sensitive Appearance: Colourless Odour: Pungent Melting Point: -45° Boiling Point: 166-168° Beilstein Reference: 507066 Merck Reference: 14,7826 UN No.: 2496 MDL No.: MFCD00009303

  • CAS Number:123-62-6
  • Formula:C₆H₁₀O₃
  • MDL Number:MFCD00009303
  • Molecular Weight:130.14 g/mol
  • Merck Index:14,7826
  • Boiling Pt:166…168 °C
  • Melting Pt:–45 °C
  • Density:1.012
  • Flash Pt:63 °C (145 °F)
  • Storage Temperature:Ambient
  • EINECS:204-638-2
  • UN:2496
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications