Propionzuuranhydride ≥97%
Glass bottle for solids, 5.000 g
Purity: 97% CAS No.: 123-62-6 Molecular Formula: C6H10O3 Formula Weight: 130.14 Flash Point: 63°(145°F) Physical Form: Clear liquid Density: 1.012 Refractive Index: 1.4040 Sensitivity: Moisture Sensitive Appearance: Colourless Odour: Pungent Melting Point: -45° Boiling Point: 166-168° Beilstein Reference: 507066 Merck Reference: 14,7826 UN No.: 2496 MDL No.: MFCD00009303
- :123-62-6
- :C₆H₁₀O₃
- :MFCD00009303
- :130.14 g/mol
- :14,7826
- :166…168 °C
- :–45 °C
- :1,012
- :63 °C (145 °F)
- :Kamertemperatuur
- :204-638-2
- :2496
- :8,III
- Verpakking:Glass bottle for solids
- VPE:5.000 g
- Cat. No.:A12955.0I



