Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Hypoxanthine ≥99%

Hypoxanthine ≥99%

Glass bottle for solids, 100 g
Catalog # A11481.22
Hypoxanthine ≥99%
Catalog # A11481.22
CAS Number:  68-94-0
Hypoxanthine ≥99%
Hypoxanthine ≥99%

Glass bottle for solids, 100 g

CAS Number:  68-94-0
Price
276,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 99% CAS No.: 68-94-0 Molecular Formula: C5H4N4O Formula Weight: 136.11 Storage Temperature: 2°C to 8°C Melting Point: 300° Beilstein Reference: 5811 Merck Reference: 14,4869 MDL No.: MFCD00005725

Synonyms:  1H-Purin-6-one, 1,9-Dihydro-6H-purin-6-one, Purin-6(1H)-one, 6-Purinone
  • CAS Number:68-94-0
  • Formula:C₅H₄N₄O
  • MDL Number:MFCD00005725
  • Molecular Weight:136.11 g/mol
  • Merck Index:14,4869
  • Melting Pt:>300 °C
  • EINECS:200-697-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications