Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

Morpholine ≥99%

Glass bottle for solids, 500 g
Catalog # A10355.36
Corrosive
Flammable
Irritant
Morpholine ≥99%
Catalog # A10355.36
CAS Number:  110-91-8
Morpholine ≥99%
Morpholine ≥99%

Glass bottle for solids, 500 g

CAS Number:  110-91-8
Price
31,80 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 99% CAS No.: 110-91-8 Molecular Formula: C4H9NO Formula Weight: 87.12 Flash Point: 35°(95°F) Physical Form: Liquid Density: 1.007 Refractive Index: 1.4540 Sensitivity: Hygroscopic Appearance: Colourless Odour: Amine-like Melting Point: -5° Boiling Point: 129° Beilstein Reference: 102549 Merck Reference: 14,6277 UN No.: 2054 MDL No.: MFCD00005972

  • CAS Number:110-91-8
  • Formula:C₄H₉NO
  • MDL Number:MFCD00005972
  • Molecular Weight:87.12 g/mol
  • Merck Index:14,6277
  • Boiling Pt:129 °C
  • Melting Pt:–5 °C
  • Density:1.007
  • Flash Pt:35 °C (95 °F)
  • Storage Temperature:Ambient
  • EINECS:203-815-1
  • UN:2054
  • ADR:8,I
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.